missing translation for 'onlineSavingsMsg'
Learn More

Olmesartan Medoxomil Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013119
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013119 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013119 Supplier Merck Emd Millipore Supplier No. PHR1851200MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 144689-63-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C29H30N6O6
MDL Number MFCD00944911
Synonym Olmesartan medoxomil
Type Certified Reference Material
Quantity 200 mg
Formula Weight 558.59
Grade Certified Reference Material