missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olive oil, pure, refined
CAS: 8001-25-0
$42.49 - $473.87
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC416530250
|
Thermo Scientific Chemicals
416530250 |
25 mL | Glass bottle |
Each for $42.49
|
|
||||
|
AC416535000
|
Thermo Scientific Chemicals
416535000 |
500 mL | Glass bottle |
Each for $140.73
|
|
||||
|
AC416530025
|
Thermo Scientific Chemicals
416530025 |
2.5 L | Glass bottle |
Each for $473.87
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 8001-25-0 | |
| MFCD00131764 |
Specifications
| 0.3 max. (mg KOH/g) | |
| 1.20 max. (1g/100ml cyclohexane) measured at the maximum between 264nm and 276nm | |
| 0.9135g/mL | |
| Authentic | |
| 1.4670 to 1.4710 | |
| 25 mL | |
| 14, 6836 | |
| Pure | |
| Viscous Liquid |
| 8001-25-0 | |
| Colorless or Green-Yellow | |
| 225°C | |
| Glass bottle | |
| MFCD00131764 | |
| 0.9135 | |
| Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C) | |
| of fatty acids: 0.1% max. (< C16), of the sterol fraction: Sterols: min. 93.0%, Cholesterol: max. 0.5%, Campesterol: max. 4.0%, delta 7-Stigmasterol: max. 0.5% | |
| Olive oil |
Safety and Handling
EINECSNumber : 232-277-0
RTECSNumber : RK4300000
TSCA : TSCA
RUO – Research Use Only