missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olive oil, pure, refined
CAS: 8001-25-0
Supplier: Thermo Scientific Chemicals 416530025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Olive oil | |
| 8001-25-0 | |
| Colorless or Green-Yellow | |
| 225°C | |
| Glass bottle | |
| MFCD00131764 | |
| 0.9135 | |
| Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C) | |
| of fatty acids: 0.1% max. (< C16), of the sterol fraction: Sterols: min. 93.0%, Cholesterol: max. 0.5%, Campesterol: max. 4.0%, delta 7-Stigmasterol: max. 0.5% |
| 0.3 max. (mg KOH/g) | |
| 1.20 max. (1g/100ml cyclohexane) measured at the maximum between 264nm and 276nm | |
| 0.9135g/mL | |
| Authentic | |
| 1.4670 to 1.4710 | |
| 2.5 L | |
| 14, 6836 | |
| Pure | |
| Viscous Liquid |
Chemical Identifiers
| 8001-25-0 | |
| MFCD00131764 |
Safety and Handling
EINECSNumber : 232-277-0
RTECSNumber : RK4300000
TSCA : TSCA
RUO – Research Use Only