missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Oligo(dT)18 Primer

Thermo Scientific™ Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.

Manufacturer:  Thermo Scientific™ SO132

 View more versions of this product

Catalog No. FERSO132



Description

Description

Thermo Scientific™ Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.

The primer is supplied as a ready-to-use, 20X concentrated aqueous solution for use in cDNA synthesis.
Specifications

Specifications

Contains 60 µg Oligo(dT)18 Primer at 100 µM.
Store at -20°C.
Oligo dT Primers
120µL (30µg)
Documents
Promotions

Promotions