missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oligo(dT)18 Primer

Catalog No. p-4074361
Change view
Click to view available options
Quantity:
60 μL
120 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSO131 60 μL
FERSO132 120 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERSO131 Supplier Thermo Scientific™ Supplier No. SO131
Only null left

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Applications
• cDNA Synthesis
• RT-qPCR
• RT-PCR

Related products
Anchored Oligo dT (Cat. No. AB1247)
Random Hexamer Primer (Cat. No. SO142)

Specifications

Product Type Primer
Content And Storage • Oligo(dT)18 Primer (60 μL at 0.5 μg/μL)

Store at –20°C.
Primer Length 18-mer
Shipping Condition Dry Ice
Concentration 0.5 μg/μL
Primer Oligo dT
Quantity 60 μL
For Use With (Application) cDNA Synthesis
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.