missing translation for 'onlineSavingsMsg'
Learn More

Olig1 Mouse anti-Human, Mouse, Rat, Clone: TH11-2G6, MilliporeSigma™

Catalog No. p-7222084
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5540I25 25 μg
MAB5540I100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB5540I25 Supplier MilliporeSigma Supplier No. MAB5540I25UG
Only null left

Mouse Monoclonal Antibody

Olig1 Monoclonal antibody specifically detects Olig1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blotting.

Specifications

Antigen Olig1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone TH11-2G6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Recombinant fusion protein from mouse Oligodendrocyte transcription factor 1 (Olig1).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less