missing translation for 'onlineSavingsMsg'
Learn More

Oleyl Acetate 60.0+%, TCI America™

Catalog No. A0934500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0934500ML 500 mL
A093425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0934500ML Supplier TCI America Supplier No. A0934500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 693-80-1
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.52
MDL Number MFCD00056326
InChI Key GYGAZRPDUOHMAF-KHPPLWFESA-N
Synonym Acetic Acid Oleyl Ester
PubChem CID 5363289
IUPAC Name (9Z)-octadec-9-en-1-yl acetate
SMILES CCCCCCCC\C=C/CCCCCCCCOC(C)=O

Specifications

Color Yellow
Boiling Point 186°C
Quantity 500 mL
Formula Weight 310.52
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Oleyl Acetate
EINECSNumber (2)-0762
TSCA No