accessibility menu, dialog, popup

UserName

H3COCH3O(Z)

CAS RN 693-80-1

IUPAC Name: (9Z)-octadec-9-en-1-yl acetate
Synonyms: oleyl acetate
Molecular Weight (g/mol): 310.52
Molecular Formula: C20H38O2
InChi Key: GYGAZRPDUOHMAF-KHPPLWFESA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results