missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oleoyl-L-alpha-lysophosphatidic acid sodium salt

Catalog No. AAJ66836MB
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66836MB 25 mg
AAJ66836LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66836MB Supplier Thermo Scientific Chemicals Supplier No. J66836MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22556-62-3 | C21H40NaO7P, C21H40NaO7P | 458.51 g/mol

It is an endogenous agonist for LPA1 and LPA2 receptors. LPA does not induce angiogenesis, but has effects on endothelial cell physiology that are similar to those of sphingosine 1-phosphate. Induces cell migration of cancer and non-cancer cells. LPA has also been described to potentiate acetylchloine receptor currents by G-protein-mediated activation of Ca2+-dependent/independent protein kinase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22556-62-3
Molecular Formula C21H40NaO7P
Synonym 1-O-9Z-Octadecenoyl-sn-glyceryl-3-phosphoric acid sodium salt; Lysophosphatidic Acid

Specifications

Quantity 25 mg
Description White to off-white
Content And Storage -30°C to -10°C
Formula Weight 458.5
Sensitivity Air and Light Sensitive, Hygroscopic
Solubility Soluble in water up to 5mg/ml with sonication
Product Type Oleoyl-L-alpha-lysophosphatidic acid sodium salt
Form Waxy solid or powder

RUO – Research Use Only