accessibility menu, dialog, popup

UserName

CAS RN 22556-62-3

IUPAC Name: sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Synonyms: sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Molecular Weight (g/mol): 458.51
Molecular Formula: C21H40NaO7P
InChi Key: XGRLSUFHELJJAB-UHFFFAOYNA-M
SMILES: [Na+].CCCCCCCCC=CCCCCCCCC(=O)OCC(O)COP(O)([O-])=O
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)