missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleic Acid 99.0+%, TCI America™
$91.13 - $330.36
Chemical Identifiers
| CAS | 112-80-1 |
|---|---|
| Molecular Formula | C18H34O2 |
| Molecular Weight (g/mol) | 282.47 |
| MDL Number | MFCD00064242 |
| InChI Key | ZQPPMHVWECSIRJ-MDZDMXLPSA-N |
| Synonym | oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid |
| PubChem CID | 445639 |
| ChEBI | CHEBI:16196 |
| IUPAC Name | (9E)-octadec-9-enoic acid |
| SMILES | CCCCCCCC\C=C\CCCCCCCC(O)=O |
Chemical Identifiers
| 112-80-1 | |
| 282.47 | |
| ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
| 445639 | |
| (9E)-octadec-9-enoic acid |
| C18H34O2 | |
| MFCD00064242 | |
| oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid | |
| CHEBI:16196 | |
| CCCCCCCC\C=C\CCCCCCCC(O)=O |
Specifications
| 112-80-1 | |
| 285°C | |
| MFCD00064242 | |
| oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid | |
| CCCCCCCC\C=C\CCCCCCCC(O)=O | |
| 282.47 | |
| CHEBI:16196 | |
| ≥99.0% (GC,T) | |
| Oleic Acid |
| Yellow | |
| C18H34O2 | |
| 5 mL | |
| ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
| (9E)-octadec-9-enoic acid | |
| 445639 | |
| 282.47 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0609&(2)-0975
RTECSNumber : RG2275000
TSCA : Yes
Recommended Storage : Refrigerator