missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid 99.0+%, TCI America™

Supplier:  TCI America O018025ML

 View more versions of this product

Catalog No. O018025ML


Only null left

Description

Description

Specifications

Specifications

Oleic Acid
Yellow
C18H34O2
25 mL
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(9E)-octadec-9-enoic acid
445639
282.47
Liquid
112-80-1
285°C
MFCD00064242
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
CCCCCCCC\C=C\CCCCCCCC(O)=O
282.47
CHEBI:16196
≥99.0% (GC,T)

Chemical Identifiers

112-80-1
282.47
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
445639
(9E)-octadec-9-enoic acid
C18H34O2
MFCD00064242
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
CHEBI:16196
CCCCCCCC\C=C\CCCCCCCC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609&(2)-0975

RTECSNumber : RG2275000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions