missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleic Acid, Purified, (cis-9-Octadecenoic Acid), Reagents
Oleic Acid, Purified, (cis-9-Octadecenoic Acid), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$171.89 - $686.29
Specifications
| Chemical Name or Material | Oleic Acid, Purified, (cis-9-Octadecenoic Acid) |
|---|---|
| CAS | 112-80-1 |
| Boiling Range | 286°C |
| Melting Point | 14°C |
| Color | Colorless to Slight Yellow |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-583
|
Labchem Inc
C2110000100C |
100 mL |
Each for $171.89
|
|
|||||
|
01-337-582
|
Labchem Inc
C21100001C |
1 L |
Each for $686.29
|
|
|||||
|
01-337-581
|
Labchem Inc
C2110000500C |
500 mL |
Each for $443.07
|
|
|||||
Specifications
| Oleic Acid, Purified, (cis-9-Octadecenoic Acid) | |
| 286°C | |
| Colorless to Slight Yellow | |
| 14°C | |
| C18H34O2 | |
| cis-9-Octadecenoic Acid | |
| Reagent |
| 112-80-1 | |
| 14°C | |
| 286°C | |
| Glass Bottle | |
| 0.895 | |
| 282.2559 | |
| Liquid |