missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleic Acid, Food Grade, Spectrum™ Chemical
O1500, 112-80-1, C18H34O2
$519.79 - $3,200.70
Chemical Identifiers
| CAS | 112-80-1 |
|---|---|
| Molecular Formula | C18H34O2 |
| Molecular Weight (g/mol) | 282.47 |
| MDL Number | MFCD00064242 |
| InChI Key | ZQPPMHVWECSIRJ-MDZDMXLPSA-N |
| IUPAC Name | (9E)-octadec-9-enoic acid |
| SMILES | CCCCCCCC\C=C\CCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604878
|
Spectrum Chemical Mfg Cor
O15001LTGL |
1 L | Amber Glass Bottle |
N/A
|
|
||||
|
18604879
|
Spectrum Chemical Mfg Cor
O15004LTGL |
4 L | Amber Glass Bottle |
N/A
|
|
||||
|
18604881
|
Spectrum Chemical Mfg Cor
O150020LT |
20 L | Amber Glass Bottle |
N/A
|
|
||||
|
18604880
|
Spectrum Chemical Mfg Cor
O1500200LTBL |
200 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Oleic Acid, Food GradeChemical Identifiers
| 112-80-1 | |
| 282.47 | |
| ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
| CCCCCCCC\C=C\CCCCCCCC(O)=O |
| C18H34O2 | |
| MFCD00064242 | |
| (9E)-octadec-9-enoic acid |
Specifications
| 112-80-1 | |
| 286.1°C | |
| C18H34O2 | |
| 1 L | |
| ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
| (9E)-octadec-9-enoic acid | |
| Food Grade |
| 1 | |
| Amber Glass Bottle | |
| MFCD00064242 | |
| 196-204 | |
| CCCCCCCC\C=C\CCCCCCCC(O)=O | |
| 282.47 | |
| Liquid |