missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, 99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215178101

 View more versions of this product

Catalog No. ICN15178101

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Oleic acid
White
C18H34O2
1 g
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(9E)-octadec-9-enoic acid
445639
282.52
Solid
112-80-1
0.89g/L
MFCD00064242
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
CCCCCCCC\C=C\CCCCCCCC(O)=O
282.47
CHEBI:16196
0.99

Chemical Identifiers

112-80-1
282.47
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
445639
(9E)-octadec-9-enoic acid
C18H34O2
MFCD00064242
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
CHEBI:16196
CCCCCCCC\C=C\CCCCCCCC(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions