missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, 99%, MP Biomedicals™

Catalog No. p-4404843
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15178101 1 g
ICN15178105 5 g
ICN15178125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15178101 Supplier MP Biomedicals Inc Supplier No. 0215178101
Only null left

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Color White
Density 0.89g/L
Quantity 1 g
Formula Weight 282.52
Percent Purity 0.99
Physical Form Solid
Chemical Name or Material Oleic acid