missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic Acid Hydrate 98.0+%, TCI America™

Catalog No. O03175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03175G 5 g
O031725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03175G Supplier TCI America Supplier No. O03175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 508-02-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.711
MDL Number MFCD00064914
InChI Key MIJYXULNPSFWEK-GTOFXWBISA-N
Synonym oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid
PubChem CID 10494
ChEBI CHEBI:37659
IUPAC Name (4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C

Specifications

Melting Point 311°C
Color White
Quantity 5 g
Formula Weight 456.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Oleanolic Acid Hydrate
RTECSNumber RK0177965
TSCA Yes