missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic Acid Hydrate 98.0+%, TCI America™

Supplier:  TCI America O031725G

Quantity 25 g

 View more versions of this product

Catalog No. O031725G

Only null left

Description

Description

Specifications

Chemical Identifiers

508-02-1
456.711
MIJYXULNPSFWEK-GTOFXWBISA-N
10494
(4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
C30H48O3
MFCD00064914
oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid
CHEBI:37659
CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C

Specifications

Oleanolic Acid Hydrate
311°C
C30H48O3
MFCD00064914
MIJYXULNPSFWEK-GTOFXWBISA-N
(4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
10494
456.71
Crystalline Powder
508-02-1
White
25 g
oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid
CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C
456.711
CHEBI:37659
≥98.0% (T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : RK0177965

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions