missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic acid, 97%

Catalog No. AAH60445MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60445MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60445MD Supplier Thermo Scientific Chemicals Supplier No. H60445MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 508-02-1 | C30H48O3 | 456.711 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 508-02-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.711
MDL Number MFCD00064914
InChI Key MIJYXULNPSFWEK-GTOFXWBISA-N
Synonym oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid
PubChem CID 10494
ChEBI CHEBI:37659
IUPAC Name (4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C

Specifications

Melting Point 300°C
Quantity 250 mg
Formula Weight 456.7
Percent Purity 97%
Chemical Name or Material Oleanolic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-081-6
RTECSNumber RK0177965
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only