missing translation for 'onlineSavingsMsg'
Learn More

Olanzapine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018682
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018682 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018682 Supplier Merck Emd Millipore Supplier No. PHR18251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Olanzapine is an atypical antipsychotic drug, widely used for the treatment of acute schizophrenia and psychotic conditions.

Specifications

CAS 132539-06-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H20N4 S
MDL Number MFCD00866702
Synonym Olanzapine; 2-Methyl-4-(4-methyl-1-piperazinyl)- 10 H-thieno[2,3-b][1,5 ]benzodiazepine
Type Certified Reference Material
Quantity 1 g
Formula Weight 312.43
Grade Certified Reference Material