Learn More
Thermo Scientific™ Okadaic acid sodium salt, 98%, high purity
Supplier: Thermo Scientific™ 328400001
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Okadaic acid sodium salt | |
| 209266-80-8 | |
| 100.0 | |
| 98% | |
| MFCD00210208 | |
| okadaic acid sodium salt, okadaic acid sodium salt from prorocentrum concavum, okadaic acid sodium salt from prorocentrum concavum hplc , film | |
| BYHIFOCTDVNQQT-GHIYGBLASA-M | |
| sodium;(2R)-3-[(2S,6R,8S,11R)-2-[(E,2R)-4-[(2S,2'R,4R,4aS,6R,8aR)-4-hydroxy-2-[(1S,3S)-1-hydroxy-3-[(2S,3R,6S)-3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl]butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl] | |
| 71308608 | |
| 98% |
| 98% | |
| 97.5 | |
| Authentic | |
| C44H67NaO13 | |
| 0.1mg | |
| Solubility in water: insoluble in water. Other solubilities: soluble in dmso | |
| CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[Na+] | |
| 827 | |
| 827 |
Chemical Identifiers
| 209266-80-8 | |
| 827 | |
| BYHIFOCTDVNQQT-GHIYGBLASA-M | |
| 71308608 | |
| CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[Na+] |
| C44H67NaO13 | |
| MFCD00210208 | |
| okadaic acid sodium salt, okadaic acid sodium salt from prorocentrum concavum, okadaic acid sodium salt from prorocentrum concavum hplc , film | |
| sodium;(2R)-3-[(2S,6R,8S,11R)-2-[(E,2R)-4-[(2S,2'R,4R,4aS,6R,8aR)-4-hydroxy-2-[(1S,3S)-1-hydroxy-3-[(2S,3R,6S)-3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl]butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl] |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO â Research Use Only