accessibility menu, dialog, popup

UserName

CAS RN 209266-80-8

CH3OOCH3CH2OOOCH3OOCH3OOOHHOH3COHOHNa(E/Z)

CAS RN 209266-80-8

IUPAC Name: sodium 2-hydroxy-3-(5-hydroxy-8-{4-[8'-hydroxy-6'-(1-hydroxy-3-{3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl}butyl)-7'-methylidene-hexahydro-3'H-spiro[oxolane-2,2'-pyrano[3,2-b]pyran]-5-yl]but-3-en-2-yl}-10-methyl-1,7-dioxaspiro[5.5]undec-10-en-2-yl)-2-methylpropanoate
Synonyms: sodium 2-hydroxy-3-(5-hydroxy-8-{4-[8'-hydroxy-6'-(1-hydroxy-3-{3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl}butyl)-7'-methylidene-hexahydrospiro[oxolane-2,2'-pyrano[3,2-b]pyran]-5-yl]but-3-en-2-yl}-10-methyl-1,7-dioxaspiro[5.5]undec-10-en-2-yl)-2-methylpropanoate
Molecular Weight (g/mol): 827
Molecular Formula: C44H67NaO13
InChi Key: BYHIFOCTDVNQQT-UHFFFAOYNA-M
SMILES: [Na+].CC(CC(O)C1OC2CCC3(CCC(O3)C=CC(C)C3CC(C)=CC4(OC(CC(C)(O)C([O-])=O)CCC4O)O3)OC2C(O)C1=C)C1OC2(CCCCO2)CCC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results