Learn More
Troemner™ OIML Weight Cases
Polycarbonate Case for OIML Precision Weight
Supplier: Troemner™ 80864289
Description
- Polycarbonate Case for OIML Precision Weight has a comfortable handle, a recessed grip for removing the lid, side latches for holding the lid in place, and a color-coded insert.
- Urethane bumpers, both inside the lid and on the bottom of the case, gently hold the weight in place.
Specifications
| Individual OIML Weight Case | |
| Polycarbonate | |
| 500g OIML weight | |
| Each |