Learn More
Parker domnick hunter™ Oil-XPlus™ Filter Elements for Gas Generators
Catalog No. 1495018
Change view
Click to view available options
Type:
Inlet Filter Element
Catalog No. Type
1495018 Inlet Filter Element
1 options
Catalog No. 1495018 Supplier Parker domnick hunter™ Supplier No. K009AO
Only null left
Replacement elements for >Oil-XPlus Compressed Air Filters used with Zero Air Generators
  • Use specially treated borosilicate glass microfibers to repel oil and water, reducing pressure drop and minimizing running costs
  • Comply with ISO 8573-1 Air Quality Class 1.3.1
  • Maximum usage: 8000 hours
Specifications
Type Inlet Filter Element
Certifications/Compliance ISO 8573-1
For Use With (Equipment) Uhp-10za and uhp-35za