missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oil Violet, TCI America™
Supplier: TCI America O00965G
Specifications
| Oil Violet | |
| 6368-72-5 | |
| MFCD00003904 | |
| VKWNTWQXVLKCSG-UHFFFAOYSA-N | |
| N-ethyl-1-[(4-phenyldiazenylphenyl)diazenyl]naphthalen-2-amine | |
| 61396 | |
| 5 g |
| 140°C | |
| C24H21N5 | |
| Fat Red 7B, Solvent Red 19, Sudan Red 7B | |
| CCNC1=C(C2=CC=CC=C2C=C1)N=NC3=CC=C(C=C3)N=NC4=CC=CC=C4 | |
| 379.467 | |
| 379.47 | |
| Crystal/Powder |
Chemical Identifiers
| 6368-72-5 | |
| 379.467 | |
| VKWNTWQXVLKCSG-UHFFFAOYSA-N | |
| 61396 | |
| CCNC1=C(C2=CC=CC=C2C=C1)N=NC3=CC=C(C=C3)N=NC4=CC=CC=C4 |
| C24H21N5 | |
| MFCD00003904 | |
| Fat Red 7B, Solvent Red 19, Sudan Red 7B | |
| N-ethyl-1-[(4-phenyldiazenylphenyl)diazenyl]naphthalen-2-amine |
Safety and Handling
EINECSNumber : (5)-3086
RTECSNumber : QM2850000
TSCA : Yes