accessibility menu, dialog, popup

UserName

H3CNHNNNN(E/Z)(E/Z)

CAS RN 6368-72-5

IUPAC Name: N-ethyl-1-{2-[4-(2-phenyldiazen-1-yl)phenyl]diazen-1-yl}naphthalen-2-amine
Synonyms: sudan red 7B
Molecular Weight (g/mol): 379.47
Molecular Formula: C24H21N5
InChi Key: VKWNTWQXVLKCSG-UHFFFAOYSA-N
SMILES: CCNC1=CC=C2C=CC=CC2=C1N=NC1=CC=C(C=C1)N=NC1=CC=CC=C1
Grade 
  • (1)
  • (1)
Quantity 
  • (2)
Color 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)