missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Oil-Selective Sorbents

Selectively remove hydrocarbons from water

Supplier:  Fisherbrand™ FBB810

 View more versions of this product

Catalog No. 172152A


Only null left

Description

Description

Multilayer fine fiber polypropylene sorbents sorb up to 25 times their weight in petrochemical-based liquids.

  • For sorbing oil and a wide variety of acids and other hazardous chemicals
  • Unaffected by temperature, humidity and pest infestation; easy to store
  • Can be cleaned and reused repeatedly

Booms

  • Polypropylene sorbents absorb oil and solvent spills on water
  • Booms available with Sock-in-Net or open netting covering designs
  • Booms float on the surface to selectively remove hydrocarbons and contain the spill
  • Hardware is included to link booms
Pads, Pillows and Rolls
  • Multilayer fine fiber material for cleaning spills of flammable material
  • Sorbent pads and rolls come in a range of sizes and can be trimmed to fit
  • Compact pillow, socks, and pads can be placed directly on spills
Particulates
  • Can be scattered on petroleum spills
  • Use particulates exactly where needed

Specifications

Chemical Identifiers

Polypropylene
Sorbing oil and a wide variety of acids and other hazardous chemicals

Specifications

70 gal.
Polypropylene
Sorbing oil and a wide variety of acids and other hazardous chemicals
10 ft.
8 in.
265 L
Oil Sorbent Boom
10 ft. x 8 in. (3.04 m x 20.3 cm)
3.04 m
20.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions