Learn More
Oil and Grease in Seawater, Whole Volume, WP Proficiency Testing Material, MilliporeSigma™ Supelco™
Description
This is a whole volume sample for the analysis of oil and grease. This sample is only suitable for gravimetric methods. The solvent for this sample is sea water.Sample preparationThis sample is ready to analyze. For calculation purposes, assume the volume of the sample to be 1 L. Rinse the bottle well with your extraction solvent including the cap. Analyze using gravimetric methods.
Specifications
Specifications
| Quantity | 250 mL |
| Concentration or Composition (by Analyte or Components) | n-Hexane Extractable Material (O&G;) 20-100 mg/L |