missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Triple Beam Balance Mass Set

Catalog No. S40037
Change view
Click to view available options
Weight (Metric):
1 x 500 g, 2 x 1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
S40037 1 x 500 g, 2 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40037 Supplier OHAUS™ Supplier No. OHS 707 EMD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with a variety of mechanical scales. OHAUS™ Triple Beam Balance Mass Set includes two 1kg and one 500g counterweights for use with compatible Triple Beam 700 series, Dial-O-Gram 1600 series and Pro series mechanical balances.

Counterweight set for use with the following mechanical scales:

  • Triple Beam 710-00
  • Triple Beam 750-S0
  • Triple Beam 760-00
  • Triple Beam Dial-O-Gram 1650-00
  • Triple Beam Pro TP2611

Specifications

Product Type Metric Attachment Weight Set
Description Weight Set, 2x1kg, 1x500g
Classification Miscellaneous
Material Cast Iron
Weight (Metric) 1 x 500 g, 2 x 1 kg
Shape Cylindrical
For Use With (Application) Calibration, counter weight
For Use With (Equipment) Triple Beam 710-00, 750-S0 and 760-00; Triple Beam Dial-O-Gram 1650-00; and Triple Beam Pro TP2611
Packaging Quantity 3 Weights/Pk.