Learn More
OHAUS™ RS232 Cable for SF42 Printers
Catalog No. 0192133
Change view
Click to view available options
RS232 Cable for SF42 printer
Catalog No.
0192133 RS232 Cable for SF42 printer
1 options
Catalog No. 0192133 Supplier OHAUS™ Supplier No. 80500571
Only null left
Connects OHAUS™ Adventure Pro, Discovery, MB Series, Ranger Series Balances to the OHAUS™ SF42 printer.
For use with Adventure Pro, Discovery, MB Series, Ranger Series Balances.
Specifications
For Use With (Equipment) SF42 Printer for Adventure Pro, Discovery, MB Series
Type RS232 Cable