missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Navigator™ XL Balance

Ultrafast stabilization in less than one second provides fast, accurate and repeatable weighing results

Supplier:  Ohaus™ 30573836

Catalog No. 01918316

Only null left
Explore available promotions

Description

Description

Two touchless sensors free up your hands for handling samples and limiting keypad wear and tear.
 
  • Superior mechanical overload protection system and integrated shipping lock
  • Withstands loads up to 400% of its rated capacity
  • Optional RS-232 or USB or ethernet interface kits with integrated cables
  • Up front illuminated level indicator
  • Removable stainless-steel weighing platform
  • Menu lockswitch
  • Integral security bracket and integral shipping lock
  • Stability indicator, overload and underload indicators, low battery indicator, auto shut-off
  • ABS plastic housing; 304 stainless steel pan
Specifications

Specifications

4 Mechanical keys, raised and with tactile feedbac
RS-232 or USB connectivity
Backlit LCD
Pound, ounce (decimal/fractional), pound: ounce (d
±2 g
NVL20000/1
≤1 sec.
10°C to 40°C
2 Years
20.4 cm
7.4 cm
28.2 cm
2 kg
100/240 V
50/60 Hz
  • Electromagnetic Compatibility: C-Tick, CE Class B, FCC Class B, ICES-003 Class B
  • Safety (AC adaptors only): C-Tick, CCC, CE, cCSAus, KTL, PSE, TUV-GS; Compliant to Directive 2002/96/EC on waste electrical and electronic equipment (WEEE); Compliant to Directive 2002/95/EC on the restriction of the use of certain hazardous substances in electrical and electronic equipment (RoHS)
  • Quality: OHAUS quality management system registered to ISO 9001:2008
20 kg
1 g
AC adapter or internal rechargeable battery
ABS Plastic, Stainless Steel
1 g
50°F to 104°F
Navigator™ XL Scale
8 in.
2.9 in.
11.1 in.
4.3 lb.
100/240 V 50/60 Hz
0.25 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions