missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Explorer™ Precision Balances, NTEP Certified

Delivers accurate results within seconds, improving operator efficiency, productivity and throughput. OHAUS Explorer Precision Balances feature a color touch display that can be separated from the weighing base.

Supplier:  OHAUS™ 83021352

 View more versions of this product

Catalog No. 01-919-242

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Full-color VGA graphic display (5.7 in. diagonal), 4-wire resistive touch screen
  • QWERTY keyboard and numeric keypad
  • Integral weigh below hook for below balance weighing application
  • Removable stainless-steel weighing platform, stability indicator, overload and underload indicators
  • Metal base, ABS top housing, stainless-steel pan, glass draftshield with flip-top door, replaceable in-use cover
  • Standard USB and R-S232 communication ports and an optional third port of either RS-232 or ethernet
  • GLP/GMP data ouput with real time clock
  • Data transfer function
  • AutoCal fully-automatic internal calibration system
  • Fast stabilization time
Specifications

Specifications

Touch screen
Bi-directional RS-232, USB
Color Graphic
Gram, milligram, kilogram, carat, ounce, troy ounc
±0.002 mg
EX1103N
1 kg
≤1.5 sec.
10°C to 30°C
9.1 in.
13 cm
35.02 cm
26 cm
7.5 kg
100/240 V
50/60 Hz
NTEP and OIML E2 or ASTM CL 1
1100 g
0.001 g
AC adapter, user manual, AutoCal™ fully-auto
ABS Plastic, Stainless Steel, Glass
<±0.001 g
3ppm/°C
50°F to 86°F
Precision Balance
23 cm
13.8 in.
10.2 in.
16.5 lb.
100/240 V 50/60 Hz
0.6 A
Class I
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions