Learn More
OHAUS™ Bench Scales Computer Data Transfer Cables
Catalog No. 0191826
Change view
Click to view available options
For Use With (Equipment):
Models B10S05, B25S05, B50S05, B100S05, B150S05
Catalog No. For Use With (Equipment)
0191826 Models B10S05, B25S05, B50S05, B100S05, B150S05
1 options
Catalog No. 0191826 Supplier OHAUS™ Supplier No. 12106964
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Connects Link Champ General-Purpose Bench Scales to IBM-PC computers via standard RS-232-C.
Permits data collection via computer.
Specifications
For Use With (Equipment) Models B10S05, B25S05, B50S05, B100S05, B150S05
Connection Type RS-232 cable, PC 9 pin
For Use With (Application) Computer data transfer
Type Cable Kit