Learn More
OHAUS™ ASTM Class 4 Individual Weight
Catalog No. 01921300
Change view
Click to view available options
Mass:
300 g
Catalog No. Mass
01921300 300 g
1 options
Catalog No. 01921300 Supplier OHAUS™ Supplier No. 80780137
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Meets ASTM Class 4 adjustment tolerances and is ideal for applications requiring a degree of accuracy above general purpose weights
Specifications
Product Type Calibration Weight
Certifications/Compliance ASTM
Classification ASTM Class 4
Material Stainless Steel
Weight (Metric) 300 g
Mass 300 g
Quantity Each
Packaging Quantity 1