missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Adventurer™ Precision Balance

Catalog No. p-4918286
Change view
Click to view available options
Model:
AX1502
AX2202
AX223
AX4201
AX4202
AX423
AX423N
AX5202
AX523
AX622
AX8201
Capacity:
220 g
420 g
520 g
620 g
1.5 kg
2.2 kg
4.2 kg
5.2 kg
8.2 kg
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Capacity
01920257 AX4201 4.2 kg
01920259 AX8201 8.2 kg
01920266 AX622 620 g
01920268 AX1502 1.5 kg
01920270 AX2202 2.2 kg
01920272 AX4202 4.2 kg
01920273 AX5202 5.2 kg
01920275 AX223 220 g
01920277 AX423 420 g
01920279 AX523 520 g
01920281 AX423N 420 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 01920257 Supplier OHAUS™ Supplier No. 30122634
Only null left

Adventurer™ Precision Balance strikes the ideal balance between inventive features and functional, uncomplicated weighing capabilities.

Delivers stable and reliable weighing results for routine weighing tasks.

  • Fast stabilization time improves productivity in the laboratory
  • AutoCal™ OHAUS automatic internal calibration system performs routine maintenance by calibrating the balance daily
  • Color touchscreen offers easy and fast operation of Adventurer applications
  • Operate and access nine application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen
  • Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves
  • In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print
  • Application modes: weighing, dynamic weighing, display hold, parts counting, density determination, totalization, percent weighing, check weighing, formulation
  • Real-time clock function keeps accurate time even during power loss and the GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements
  • Cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances
  • When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit; simply increase your sample weight to assure that your results are up to your standards
  • Some models have a space-saving draftshield designed to improve user experience and accessibility.
  • Additional features include: RS232 interface, integral weigh below hook, full housing in-use cover, removable stainless steel pan, die-cast metal bottom housing, security bracket, illuminated up-front level indicator, four adjustable feet, software lockout menus, stability indicator, software overload/underload indicators, user selectable environmental settings, audible indicator, user selectable brightness settings, auto dim, touchscreen calibration, auto tare, user selectable operating language (nine)

Dual USB Ports

  • A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance
  • A second USB device is located at the rear of the balance that can be used to connect the balance to a PC
  • The connectivity options help meet traceability requirements in traditional installations

Compliance

Metrology: OIML R76, EN 45501 (Class I, nmax 320000, Class II, nmax 52000), Product Safety: EN 61010-1, IEC 61010-1, Electromagnetic Compatibility: IEC 61326-1, EN 61326-1 (Emissions Class B, Immunity Industrial Requirements), GLP/GMP Compliant

Specifications

Certifications/Compliance Metrology: OIML R76, EN 45501 (Class I, nmax 320000, Class II, nmax 52000), Product Safety: EN 61010-1, IEC 61010-1, Electromagnetic Compatibility: IEC 61326-1, EN 61326-1 (Emissions Class B, Immunity Industrial Requirements), GLP/GMP Compliant
Connection Type Dual USB ports
Capacity 4.2 kg
Display Type Color Touchscreen
Readability 0.1 g
For Use With (Application) Weighing, Parts Counting, Percent Weighing, Check
Linearity ±0.2 g
Material Metal Base, ABS Top Housing, Stainless Steel Pan
Model AX4201
Repeatability ±0.1 g
Stabilization Time ≤1.5 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
Type Precision Balance
Width (English) Pan 7.7 in.
Width (Metric) Pan 195 mm
Weight (English) 10.2 lb.
Weight (Metric) 4.6 kg
Electrical Requirements AC Adapter Input: 100-240V 0.3 A 50-60 Hz, AC Adapter
Show More Show Less