missing translation for 'onlineSavingsMsg'
Learn More

Ofloxacin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015624
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015624 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015624 Supplier Merck Emd Millipore Supplier No. PHR11681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Ofloxacin is an orally administered antimicrobial drug that is classified under the group of fluoroquinolones. It has high potency and shows a broad-spectrum antibacterial performance, as well as short selection of resistant bacteria.

Specifications

CAS 82419-36-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C18H20 FN3O4
MDL Number MFCD00226105
Type Certified Reference Material
Quantity 1 g
Formula Weight 361.37
Grade Certified Reference Material