missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Pellets, NF, 95-100.5%, Spectrum™ Chemical

Catalog No. 18606580
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606580 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606580 Supplier Spectrum Chemical Mfg Cor Supplier No. SO170500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO170, 1310-73-2, NaOH

Spectrum™ Chemical Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95 to 100.5%
Grade NF
Packaging Poly Bottle