missing translation for 'onlineSavingsMsg'
Learn More

Octyltrichlorosilane, 97%

Catalog No. p-3758591
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC216501000 100 g
AC216505000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC216501000 Supplier Thermo Scientific Chemicals Supplier No. 216501000
Only null left

CAS: 5283-66-9 | C8H17Cl3Si | 247.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5283-66-9
MDL Number MFCD00000488
InChI Key RCHUVCPBWWSUMC-UHFFFAOYSA-N
Synonym trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
PubChem CID 21354
IUPAC Name trichloro(octyl)silane
SMILES CCCCCCCC[Si](Cl)(Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 233°C (731.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H17Cl3Si
Refractive Index 1.4470 to 1.449
Linear Formula CH3(CH2)7SiCl3
Quantity 100 g
Beilstein 04, III, 1907
Specific Gravity 1.07
Solubility Information Solubility in water: reacts
Molecular Weight (g/mol) 247.67
Viscosity 1.9 mPa.s (25°C)
Formula Weight 247.67
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Octyltrichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 226-112-1

RUO – Research Use Only