accessibility menu, dialog, popup

UserName

H3CSiClClCl

CAS RN 5283-66-9

IUPAC Name: trichloro(octyl)silane
Synonyms: silane, trichlorooctyl-
Molecular Weight (g/mol): 247.66
Molecular Formula: C8H17Cl3Si
InChi Key: RCHUVCPBWWSUMC-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)