missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Octyl Sepharose™ 4 Fast Flow Chromatography Media

Catalog No. 45002935
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002935 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002935 Supplier Cytiva Supplier No. 17094602

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins

  • HIC ligands are coupled to monosacharide units via their corresponding glycidylethers, giving matrices without charges and stable etherbonds between ligands and agarose
  • Octyl derivative adds different and complementary selectivity to Sepharose Fast Flow HIC product family
  • Resin based on cross-linked 4% agarose
  • Supported for industrial applications and well established in approved processes
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Quantity 200 mL
For Use With (Application) For Capture and Intermediate PurifiCatIon of Larger Proteins
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 30% Isopropanol, 70% Ethanol, 6 M Guanidine-Hydrochloride, 30% Acetonitrile, 1 mM HCl
Storage Requirements 4°C to 30°C
Product Type Octyl Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Octyl
Show More Show Less