missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octyl acetate, 99+%
CAS: 112-14-1 | C10H20O2 | 172.27 g/mol
Chemical Identifiers
| CAS | 112-14-1 |
|---|---|
| Molecular Formula | C10H20O2 |
| Molecular Weight (g/mol) | 172.27 |
| MDL Number | MFCD00009562 |
| InChI Key | YLYBTZIQSIBWLI-UHFFFAOYSA-N |
| Synonym | n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8 |
| PubChem CID | 8164 |
| ChEBI | CHEBI:87495 |
| IUPAC Name | octyl acetate |
| SMILES | CCCCCCCCOC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC310631000
|
N/A | 100 g | Glass bottle |
N/A
|
|
||||
|
AC310635000
|
N/A | 500 g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C10H20O2 | |
| MFCD00009562 | |
| n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8 | |
| CHEBI:87495 | |
| CCCCCCCCOC(C)=O |
Specifications
| 112-14-1 | |
| 100.0 | |
| 0.8680g/mL | |
| 86°C | |
| 99+% | |
| 1.4175 to 1.4195 | |
| MFCD00009562 | |
| n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8 | |
| CCCCCCCCOC(C)=O | |
| 172.27 | |
| CHEBI:87495 | |
| 99+% | |
| Octyl acetate, 99+% |
| 99.0 | |
| Colorless | |
| 211.0°C | |
| Authentic | |
| C10H20O2 | |
| CH3CO2(CH2)7CH3 | |
| 0.868 | |
| YLYBTZIQSIBWLI-UHFFFAOYSA-N | |
| octyl acetate | |
| 8164 | |
| 172.27 | |
| Liquid |
Safety and Handling
EINECSNumber : 203-939-6
RTECSNumber : AJ1400000
TSCA : TSCA
RUO – Research Use Only