missing translation for 'onlineSavingsMsg'
Learn More

Octyl acetate, 99+%

Catalog No. AC310631000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310631000 100 g Glass bottle
AC310635000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310631000 Supplier Thermo Scientific Chemicals Supplier No. 310631000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-14-1 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-14-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009562
InChI Key YLYBTZIQSIBWLI-UHFFFAOYSA-N
Synonym n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
PubChem CID 8164
ChEBI CHEBI:87495
IUPAC Name octyl acetate
SMILES CCCCCCCCOC(C)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.8680g/mL
Boiling Point 211.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4175 to 1.4195
Linear Formula CH3CO2(CH2)7CH3
Quantity 100 g
Specific Gravity 0.868
Formula Weight 172.27
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Octyl acetate, 99+%
Show More Show Less
Hazard Category
EINECSNumber 203-939-6
RTECSNumber AJ1400000
TSCA TSCA

RUO – Research Use Only