missing translation for 'onlineSavingsMsg'
Learn More

Octreotide Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015557
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015557 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015557 Supplier Merck Emd Millipore Supplier No. PHR188010MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 79517-01-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C49H66N10O10S2 · xC2H4O2
Synonym Octreotide acetate salt
Type Certified Reference Material
Quantity 10 mg
Formula Weight 1019.24 (free base basis)
Grade Certified Reference Material