missing translation for 'onlineSavingsMsg'
Learn More

(±)-Octopamine hydrochloride, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015484
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015484 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015484 Supplier Merck Emd Millipore Supplier No. 68631100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 770-05-8
Physical Form Neat
Linear Formula HOC6H4 CH(CH2 NH2)OH · HCl
MDL Number MFCD00012881
Melting Point 170°C
Synonym alpha-(Aminomethyl)-4-hydroxybenzyl alcohol hydrochloride; (+/-)-1-(4-Hydroxyphenyl)-2-amino­ethanol hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 189.64
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less