missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octocrylene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 6197-30-4 |
| Density | 1.051 g/mL (at 25°C) |
| Flash Point | 234°C - closed cup |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
| Physical Form | Neat |
| Linear Formula | (C6H5)2 C=C(CN)CO2 CH2 CH(C2H5)(CH2)3 CH3 |
| MDL Number | MFCD00059260 |
| Melting Point | -10°C |
| Boiling Point | 218°C (1.5 mmHg) |
| Synonym | 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate |
| Show More |