missing translation for 'onlineSavingsMsg'
Learn More

Octocrylene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014915
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014915 10 g
111014914 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014915 Supplier Merck Emd Millipore Supplier No. PHR108310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 6197-30-4
Density 1.051 g/mL (at 25°C)
Flash Point 234°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (C6H5)2 C=C(CN)CO2 CH2 CH(C2H5)(CH2)3 CH3
MDL Number MFCD00059260
Melting Point -10°C
Boiling Point 218°C (1.5 mmHg)
Synonym 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate
Type Certified Reference Material
Quantity 10 g
Formula Weight 361.48
Grade Certified Reference Material
Show More Show Less