missing translation for 'onlineSavingsMsg'
Learn More

Octocrylene, 95%, Spectrum™ Chemical

Catalog No. 18604873
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604873 50 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604873 Supplier Spectrum Chemical Mfg Cor Supplier No. O300150ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

O3001, 6197-30-4, C24H27NO2

Spectrum™ Chemical Octocrylene is an organic compound known to be a viscous oily liquid. It is used as an ingredient in some cosmetics and sunscreens. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 6197-30-4
Molecular Formula C24H27NO2
Molecular Weight (g/mol) 361.49
MDL Number MFCD00059260
InChI Key FMJSMJQBSVNSBF-UHFFFAOYNA-N
IUPAC Name 2-ethylhexyl 2-cyano-3,3-diphenylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(C#N)=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Max % 1
Boiling Point 218°C
Packaging Amber Glass Bottle
Quantity 50 mL
Percent Purity 95%
Grade Reagent
Physical Form Liquid