missing translation for 'onlineSavingsMsg'
Learn More

Octisalate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012700
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012700 1 g
111012701 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012700 Supplier Merck Emd Millipore Supplier No. PHR10811G
Octisalate is a sunscreen agent that acts as a UV filter for UVB (280-315 nm) radiation.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 118-60-5
Density 1.014 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (HO)C6H4 CO2 CH2 CH(C2H5)(CH2)3 CH3
MDL Number MFCD00053300
Boiling Point 189°C to 190°C (21 mmHg)
Synonym 2-Ethylhexyl salicylate; Octisalate; Octyl salicylate
Type Certified Reference Material
Quantity 1 g
Formula Weight 250.33
Grade Certified Reference Material
Show More Show Less