missing translation for 'onlineSavingsMsg'
Learn More

Octinoxate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014567
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014567 10 g
111014566 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014567 Supplier Merck Emd Millipore Supplier No. PHR108010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Octinoxate belongs to a category of compounds called 'organic UV-filters (UVFs)′ which constitute an active ingredient of a variety of products, such as cosmetics, sunscreens and lip balms. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5466-77-3
Flash Point 193°C
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C18H26O3
MDL Number MFCD00072582
Synonym 2-Ethylhexyl 4-methoxycinnamate; 4-Methoxycinnamic acid 2-ethylhexyl ester; OMC; Octyl methoxycinnamate
Type Certified Reference Material
Quantity 10 g
Formula Weight 290.40
Grade Certified Reference Material
Show More Show Less