missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octenidine Dihydrochloride 98.0+%, TCI America™
$50.33 - $201.32
Chemical Identifiers
| CAS | 70775-75-6 |
|---|---|
| Molecular Formula | C36H62N4 |
| MDL Number | MFCD01938808 |
Chemical Identifiers
| 70775-75-6 | |
| C36H62N4 | |
| MFCD01938808 |
Specifications
| 70775-75-6 | |
| White-Yellow | |
| MFCD01938808 | |
| 623.84 | |
| Crystalline Powder |
| 214°C | |
| C36H62N4 | |
| 1 g | |
| ≥98.0% (HPLC,N) | |
| Octenidine Dihydrochloride |
Safety and Handling
TSCA : No